Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012872.2 | NP_001012890.1 | 2 - 3 | 309 | 119 | |
XM_015278047.2 | XP_015133533.1 | 3 - 4 | 521 | 119 | ||
XM_015278048.2 | XP_015133534.1 | 3 - 4 | 518 | 119 | ||
XM_015278049.2 | XP_015133535.1 | 2 - 3 | 360 | 119 | ||
XM_015278050.2 | XP_015133536.1 | 2 - 3 | 305 | 119 | ||
XM_015278051.2 | XP_015133537.1 | 2 - 3 | 305 | 119 | ||
XM_025148641.1 | XP_025004409.1 | 2 - 3 | 165 | 119 | ||
XM_025148642.1 | XP_025004410.1 | 2 - 3 | 327 | 119 | ||
GenBank mRNA | AJ719997.1 | - | 2 - 3 | 310 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |