Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030885.2 | NP_001026056.2 | 2 - 3 | 363 | 101 | |
GenBank mRNA | AJ719734.1 | - | 2 - 3 | 361 | 101 | |
BU299154.1 | - | 2 - 3 | 363 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |