Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001858.2 | NP_001001858.1 | 3 - 4 | 416 | 81 | |
XM_015297603.2 | XP_015153089.1 | 4 - 5 | 494 | 81 | ||
XM_015297606.2 | XP_015153092.1 | 4 - 5 | 482 | 81 | ||
XM_025142959.1 | XP_024998727.1 | 5 - 6 | 1006 | 81 | ||
XM_025142960.1 | XP_024998728.1 | 5 - 6 | 808 | 81 | ||
XM_025142961.1 | XP_024998729.1 | 4 - 5 | 813 | 81 | ||
XM_025142962.1 | XP_024998730.1 | 4 - 5 | 488 | 81 | ||
GenBank mRNA | BX930102.2 | - | 4 - 5 | 287 | 81 | |
BX934108.1 | - | 3 - 4 | 292 | 81 | ||
CR353373.1 | - | 3 - 4 | 369 | 81 | ||
CR354193.1 | - | 4 - 5 | 392 | 81 | ||
X65458.1 | - | 417 | 81 | |||
X67840.1 | - | 417 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |