Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004408.2 | NP_001004408.1 | 10 - 11 | 1392 | 77 | |
GenBank mRNA | D89547.1 | - | 10 - 11 | 1393 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |