Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205009.2 | NP_990340.2 | 4 - 5 | 977 | 94 | |
GenBank mRNA | AB016281.1 | - | 4 - 5 | 977 | 94 | |
BX259386.3 | - | 1 - 2 | 173 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |