Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205120.1 | NP_990451.1 | 10 - 11 | 1210 | 146 | |
XM_015296833.2 | XP_015152319.2 | 10 - 11 | 1275 | 146 | ||
XM_015296834.2 | XP_015152320.2 | 10 - 11 | 1206 | 146 | ||
XM_025142487.1 | XP_024998255.1 | 10 - 11 | 1213 | 146 | ||
GenBank mRNA | D37900.1 | - | 10 - 11 | 1210 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |