Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031160.1 | NP_001026331.1 | 5 - 6 | 691 | 80 | |
XM_015285719.2 | XP_015141205.1 | 5 - 6 | 676 | 80 | ||
XM_015285720.2 | XP_015141206.1 | 5 - 6 | 785 | 80 | ||
XM_015285721.2 | XP_015141207.1 | 5 - 6 | 680 | 80 | ||
XM_025149698.1 | XP_025005466.1 | 5 - 6 | 987 | 80 | ||
GenBank mRNA | AJ720429.1 | - | 5 - 6 | 691 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |