Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033869.2 | NP_001029041.1 | 8 - 9 | 881 | 93 | |
XM_015294017.2 | XP_015149503.1 | 8 - 9 | 959 | 93 | ||
XM_025154858.1 | XP_025010626.1 | 8 - 9 | 802 | 93 | ||
GenBank mRNA | CR390115.1 | - | 8 - 9 | 882 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |