Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039453.1 | NP_001034542.1 | 3 - 4 | 636 | 71 | |
GenBank mRNA | AM183252.1 | - | 3 - 4 | 636 | 71 | |
BX931084.1 | - | 3 - 4 | 702 | 71 | ||
BX933585.1 | - | 3 - 4 | 700 | 71 | ||
BX934345.1 | - | 3 - 4 | 701 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |