Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080890.1 | NP_001074359.1 | 1 - 2 | 469 | 119 | |
XM_015281592.2 | XP_015137078.1 | 1 - 2 | 535 | 119 | ||
GenBank mRNA | AY640308.1 | - | 1 - 2 | 469 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |