Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204316.3 | NP_989647.3 | 821 | 101 | ||
GenBank mRNA | S78192.1 | - | 821 | 101 | ||
U28380.1 | - | 821 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Gallus gallus |