Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006177.1 | NP_001006177.1 | 4 - 5 | 421 | 69 | |
XM_025148593.1 | XP_025004361.1 | 4 - 5 | 474 | 69 | ||
GenBank mRNA | AJ720833.1 | - | 4 - 5 | 421 | 69 | |
CR389678.1 | - | 4 - 5 | 420 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |