Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF039851.1 | - | 13 - 14 | 1677 | 70 | |
AK287031.1 | - | 22 - 23 | 3501 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |