Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK245933.1 | - | 2 - 3 | 337 | 64 | |
EU019565.1 | - | 2 - 3 | 314 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |