Search Thermo Fisher Scientific
Gene Symbol: MAP2K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005257515.2 | XP_005257572.1 | 12 | 12022 | 83 | |
XM_005257516.2 | XP_005257573.1 | 12 | 11895 | 83 | ||
XM_006721975.3 | XP_006722038.1 | 12 | 11923 | 83 | ||
XM_011525025.2 | XP_011523327.1 | 12 | 12309 | 83 | ||
XM_011525026.2 | XP_011523328.1 | 12 | 11770 | 83 | ||
XM_011525027.2 | XP_011523329.1 | 12 | 11685 | 83 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP binding cassette subfamily A member 5 | ABCA5 |
ABC13, EST90625 |
-214.5kb | Anti-sense | Chr. 17 - 69244435 - 69327182 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1483 | LINC01483 | 40.1kb | Sense | Chr. 17 - 69593987 - 69903000 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |