Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139459.1 | NP_001132931.1 | 7 - 8 | 1347 | 89 | |
NM_152609.2 | NP_689822.2 | 7 - 8 | 1221 | 89 | ||
XM_005273081.3 | XP_005273138.2 | 7 - 8 | 1006 | 89 | ||
XM_005273083.4 | XP_005273140.1 | 6 - 7 | 924 | 89 | ||
XM_011544110.2 | XP_011542412.1 | 7 - 8 | 1072 | 89 | ||
XM_011544111.1 | XP_011542413.1 | 7 - 8 | 1243 | 89 | ||
XM_011544112.1 | XP_011542414.1 | 6 - 7 | 790 | 89 | ||
XM_011544113.1 | XP_011542415.1 | 6 - 7 | 915 | 89 | ||
XM_011544114.2 | XP_011542416.1 | 6 - 7 | 630 | 89 | ||
XM_017000487.1 | XP_016855976.1 | 8 - 9 | 1261 | 89 | ||
XM_017000488.1 | XP_016855977.1 | 7 - 8 | 1004 | 89 | ||
XM_017000489.1 | XP_016855978.1 | 7 - 8 | 1071 | 89 | ||
GenBank mRNA | AK056563.1 | - | 7 - 8 | 1054 | 89 | |
AK074142.1 | - | 7 - 8 | 985 | 89 | ||
AL834246.2 | - | 7 - 8 | 1055 | 89 | ||
BC010228.2 | - | 7 - 8 | 1223 | 4151228 | 89 | |
BC036200.1 | - | 6 - 7 | 1576 | 89 | ||
BX647629.1 | - | 7 - 8 | 1006 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |