Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015261.2 | NP_056076.1 | 12 - 13 | 2125 | 64 | |
XM_011542719.2 | XP_011541021.1 | 11 - 12 | 1386 | 64 | ||
GenBank mRNA | AK124878.1 | - | 12 - 13 | 2125 | 64 | |
BC098398.1 | - | 12 - 13 | 1584 | 30528692 | 64 | |
BC144524.1 | - | 12 - 13 | 1809 | 64 | ||
D29954.1 | - | 12 - 13 | 1547 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |