Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173600.2 | NP_775871.2 | 24728 | 122 | ||
GenBank mRNA | AK093065.1 | - | 31 - 32 | 1721 | 122 | |
AY236870.1 | - | 30 - 31 | 1626 | 122 | ||
HM801842.1 | - | 24728 | 122 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat kinase 2 | LRRK2 |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
-196.3kb | Sense | Chr. 12 - 40224895 - 40369285 on NCBI Build GRCh38 |
Downstream | contactin 1 | CNTN1 |
F3, GP135, MYPCN |
126.3kb | Sense | Chr. 12 - 40692439 - 41072412 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |