Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009548.hg.1 |
TC08000701.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009548.hg.1 |
ENST00000305949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009548.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000519188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000519188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000519188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000456899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000519188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000519188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000519188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000522449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
ENST00000523373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012349.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800186048.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800186048.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800186048.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800186048.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800186048.hg.1 |
TC0800012349.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800186049.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800186049.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800186049.hg.1 |
ENST00000305949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800186049.hg.1 |
NM_022045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800186049.hg.1 |
TC0800012349.hg.1 |
Clariom D TC |