Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SYT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252065.1 | NP_001238994.1 | 6 - 7 | 896 | 60 | |
NM_001300773.1 | NP_001287702.1 | 6 - 7 | 803 | 60 | ||
NM_004200.3 | NP_004191.2 | 5 - 6 | 671 | 60 | ||
XM_005274383.4 | XP_005274440.1 | 9 - 10 | 1558 | 60 | ||
XM_005274384.2 | XP_005274441.1 | 8 - 9 | 1562 | 60 | ||
XM_005274385.4 | XP_005274442.1 | 8 - 9 | 1328 | 60 | ||
XM_005274387.4 | XP_005274444.1 | 3 - 4 | 1595 | 60 | ||
XM_005274390.4 | XP_005274447.1 | 2 - 3 | 634 | 60 | ||
XM_006718736.3 | XP_006718799.1 | 2 - 3 | 605 | 60 | ||
XM_011545335.2 | XP_011543637.1 | 9 - 10 | 1524 | 60 | ||
XM_011545336.2 | XP_011543638.1 | 8 - 9 | 1399 | 60 | ||
XM_011545337.2 | XP_011543639.1 | 8 - 9 | 1391 | 60 | ||
XM_011545338.2 | XP_011543640.1 | 8 - 9 | 1296 | 60 | ||
XM_011545339.2 | XP_011543641.1 | 7 - 8 | 1023 | 60 | ||
XM_011545340.2 | XP_011543642.1 | 6 - 7 | 889 | 60 | ||
XM_011545341.2 | XP_011543643.1 | 6 - 7 | 793 | 60 | ||
XM_011545343.2 | XP_011543645.1 | 5 - 6 | 657 | 60 | ||
GenBank mRNA | AF038535.1 | - | 5 - 6 | 664 | 60 | |
BC125170.1 | - | 5 - 6 | 629 | 40121712 | 60 | |
BM679451.1 | - | 2 - 3 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |