Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000393.3 | NP_000384.2 | 47 - 48 | 3635 | 98 | |
XM_011510573.2 | XP_011508875.1 | 47 - 48 | 3417 | 98 | ||
GenBank mRNA | AB209045.1 | - | 47 - 48 | 3497 | 98 | |
BC043613.1 | - | 9 - 10 | 730 | 98 | ||
BC086874.1 | - | 31 - 32 | 2624 | 30708812 | 98 | |
M10956.1 | - | 5 - 6 | 98 | |||
X04758.1 | - | 29 - 30 | 98 | |||
Y14690.1 | - | 47 - 48 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |