Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000468102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000468102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
ENST00000468102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005477.hg.1 |
TC06000513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005477.hg.1 |
ENST00000316637.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005477.hg.1 |
ENST00000394658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005477.hg.1 |
ENST00000542066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005477.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005478.hg.1 |
TC06000513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005478.hg.1 |
ENST00000316637.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005478.hg.1 |
ENST00000394658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005478.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000468102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000468102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
ENST00000468102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007791.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154010.hg.1 |
TC0600007791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
ENST00000316637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
ENST00000444278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
NM_022047.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154011.hg.1 |
TC0600007791.hg.1 |
Clariom D TC |