Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015631.5 | NP_056446.4 | 7 - 8 | 1131 | 151 | |
XM_011539627.1 | XP_011537929.1 | 7 - 8 | 973 | 151 | ||
XM_011539628.1 | XP_011537930.1 | 7 - 8 | 973 | 151 | ||
GenBank mRNA | AK075357.1 | - | 7 - 8 | 917 | 151 | |
AL050022.1 | - | 7 - 8 | 915 | 151 | ||
AY359022.1 | - | 7 - 8 | 895 | 151 | ||
BC009494.2 | - | 4 - 5 | 291 | 151 | ||
BC061582.1 | - | 7 - 8 | 802 | 151 | ||
BC068449.1 | - | 7 - 8 | 1131 | 5298627 | 151 | |
DQ278872.1 | - | 7 - 8 | 1131 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |