Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF447873.1 | - | 3 - 4 | 2584 | 224 | |
AJ617479.1 | - | 16 - 17 | 2005 | 133 | ||
AK055844.1 | - | 17 - 18 | 1955 | 224 | ||
AL136746.1 | - | 16 - 17 | 2006 | 133 | ||
EU392499.1 | - | 1 - 2 | 79 | 224 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |