Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000345990.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000360683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000361991.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000510842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000511130.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000512718.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000515322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000521364.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001031812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001044723.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001270572.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001270573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001270574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_004384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000345990.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000360683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000361991.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000510842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000511130.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000512718.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000515322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
ENST00000521364.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001031812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001044723.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001270572.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001270573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_001270574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
NM_004384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158343.hg.1 |
TC0500008494.hg.1 |
Clariom D TC |