Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206567.1 | NP_001193496.1 | 7 - 8 | 1619 | 98 | |
XM_005245127.3 | XP_005245184.1 | 9 - 10 | 2104 | 98 | ||
GenBank mRNA | AK296228.1 | - | 7 - 8 | 1502 | 98 | |
DA945454.1 | - | 2 - 3 | 191 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons |