Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204197.1 | NP_001191126.1 | 9 - 10 | 1121 | 81 | |
NM_015678.4 | NP_056493.3 | 49 - 50 | 8151 | 81 | ||
XM_005266346.4 | XP_005266403.1 | 49 - 50 | 8151 | 81 | ||
XM_006719805.3 | XP_006719868.1 | 48 - 49 | 8142 | 81 | ||
XM_006719806.3 | XP_006719869.1 | 48 - 49 | 8142 | 81 | ||
XM_017020545.1 | XP_016876034.1 | 49 - 50 | 8133 | 81 | ||
GenBank mRNA | AB046764.1 | - | 16 - 17 | 1863 | 81 | |
AF072371.1 | - | 3 - 4 | 338 | 81 | ||
AF467288.1 | - | 50 - 51 | 7823 | 81 | ||
AK091469.1 | - | 20 - 21 | 2310 | 81 | ||
AK122888.1 | - | 19 - 20 | 2664 | 81 | ||
AK127791.1 | - | 26 - 27 | 3502 | 81 | ||
AK294737.1 | - | 9 - 10 | 1121 | 81 | ||
AK297773.1 | - | 10 - 11 | 1219 | 81 | ||
BX640760.1 | - | 25 - 26 | 3481 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |