Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011804.hg.1 |
ENST00000328313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011804.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011804.hg.1 |
TC17000910.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011803.hg.1 |
ENST00000328313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011803.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011803.hg.1 |
TC17000910.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168660.hg.1 |
TC1700009092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168663.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168663.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168663.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168663.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168663.hg.1 |
TC1700009092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000328313.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
ENST00000576512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009092.hg.1 |
NM_178543.4 |
Refseq |