Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002927.4 | NP_002918.1 | 1 - 2 | 181 | 114 | |
GenBank mRNA | AF030107.1 | - | 1 - 2 | 189 | 114 | |
BC056866.1 | - | 1 - 2 | 180 | 4419604 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |