Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031723.3 | NP_001026893.1 | 4 - 5 | 791 | 147 | |
NM_001278310.1 | NP_001265239.1 | 3 - 4 | 619 | 147 | ||
XM_011532262.2 | XP_011530564.1 | 6 - 7 | 675 | 147 | ||
XM_011532263.2 | XP_011530565.1 | 4 - 5 | 1175 | 147 | ||
XM_011532265.2 | XP_011530567.1 | 3 - 4 | 1119 | 147 | ||
XM_017008628.1 | XP_016864117.1 | 6 - 7 | 857 | 147 | ||
GenBank mRNA | AK024343.1 | - | 2 - 3 | 318 | 147 | |
AL833076.2 | - | 3 - 4 | 2663 | 147 | ||
AL833412.1 | - | 3 - 4 | 481 | 147 | ||
AY186740.1 | - | 6 - 7 | 670 | 147 | ||
AY358272.1 | - | 4 - 5 | 645 | 147 | ||
BC022248.1 | - | 4 - 5 | 660 | 4771526 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |