Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019352.hg.1 |
ENST00000269577.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019352.hg.1 |
ENST00000348049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019352.hg.1 |
ENST00000357601.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019352.hg.1 |
ENST00000423485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019352.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019352.hg.1 |
TC17001462.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000577706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
ENST00000581055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010618.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
ENST00000423485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
ENST00000578412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
NM_001067.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183699.hg.1 |
TC1700010618.hg.1 |
Clariom D TC |