Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005258070.4 | XP_005258127.1 | 9 | 6372 | 80 | |
XM_005258072.2 | XP_005258129.1 | 9 | 6159 | 80 | ||
XM_017025374.1 | XP_016880863.1 | 8 | 6345 | 80 | ||
XM_017025375.1 | XP_016880864.1 | 8 | 6104 | 80 | ||
XM_017025376.1 | XP_016880865.1 | 8 | 6086 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |