Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017934.5 | NP_060404.3 | 34 - 35 | 4132 | 103 | |
XM_005248729.4 | XP_005248786.1 | 34 - 35 | 4129 | 103 | ||
XM_011535918.2 | XP_011534220.1 | 31 - 32 | 3788 | 103 | ||
XM_017010989.1 | XP_016866478.1 | 36 - 37 | 4274 | 103 | ||
XM_017010990.1 | XP_016866479.1 | 33 - 34 | 3872 | 103 | ||
GenBank mRNA | AF310250.1 | - | 11 - 12 | 1139 | 103 | |
AJ303102.1 | - | 34 - 35 | 3919 | 103 | ||
AK000712.1 | - | 7 - 8 | 762 | 103 | ||
AK075124.1 | - | 3 - 4 | 219 | 103 | ||
AL161957.1 | - | 11 - 12 | 3986 | 103 | ||
BC008909.2 | - | 9 - 10 | 860 | 103 | ||
BC064611.1 | - | 6 - 7 | 601 | 103 | ||
BC081569.1 | - | 12 - 13 | 1175 | 103 | ||
BC137488.1 | - | 34 - 35 | 4012 | 9021488 | 103 | |
DQ924532.1 | - | 34 - 35 | 3906 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |