Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014997.3 | NP_055812.1 | 8 - 9 | 1120 | 75 | |
XM_005250233.4 | XP_005250290.1 | 7 - 8 | 1009 | 75 | ||
GenBank mRNA | BC009977.2 | - | 6 - 7 | 533 | 75 | |
BC036464.2 | - | 8 - 9 | 1102 | 4828273 | 75 | |
BC044884.1 | - | 8 - 9 | 1122 | 5273087 | 75 | |
D87454.1 | - | 8 - 9 | 862 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |