Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126123.3 | NP_001119595.1 | 2 - 3 | 272 | 66 | |
XM_017025834.1 | XP_016881323.1 | 2 - 3 | 251 | 66 | ||
XM_017025835.1 | XP_016881324.1 | 2 - 3 | 260 | 66 | ||
GenBank mRNA | AK309035.1 | - | 2 - 3 | 260 | 66 | |
BQ645002.1 | - | 292 | 66 | |||
X67098.1 | - | 2 - 3 | 133 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |