Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SEMA6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178061.1 | NP_001171532.1 | 18 - 19 | 2096 | 147 | |
NM_001178062.1 | NP_001171533.1 | 17 - 18 | 1976 | 147 | ||
XM_005244835.3 | XP_005244892.1 | 18 - 19 | 2100 | 147 | ||
XM_017000075.1 | XP_016855564.1 | 18 - 19 | 2507 | 147 | ||
XM_017000076.1 | XP_016855565.1 | 18 - 19 | 2517 | 147 | ||
XM_017000077.1 | XP_016855566.1 | 19 - 20 | 3429 | 147 | ||
XM_017000080.1 | XP_016855569.1 | 16 - 17 | 2976 | 147 | ||
XM_017000081.1 | XP_016855570.1 | 18 - 19 | 3309 | 147 | ||
XM_017000082.1 | XP_016855571.1 | 11 - 12 | 1208 | 147 | ||
GenBank mRNA | AF339153.1 | - | 17 - 18 | 1769 | 147 | |
AF339154.1 | - | 18 - 19 | 1889 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |