Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010936.hg.1 |
TC01000674.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010936.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010936.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159929.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159929.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159929.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159929.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159929.hg.1 |
TC0100008416.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |