Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135862.2 | NP_001129334.1 | 9 - 10 | 991 | 83 | |
NM_001242450.1 | NP_001229379.1 | 8 - 9 | 1031 | 83 | ||
NM_001284296.1 | NP_001271225.1 | 9 - 10 | 1077 | 83 | ||
NM_138415.4 | NP_612424.1 | 8 - 9 | 1162 | 83 | ||
XM_006724122.1 | XP_006724185.1 | 8 - 9 | 1171 | 83 | ||
GenBank mRNA | AK091480.1 | - | 9 - 10 | 1077 | 83 | |
AK092243.1 | - | 9 - 10 | 991 | 83 | ||
AK095777.1 | - | 2 - 3 | 143 | 83 | ||
AK297317.1 | - | 8 - 9 | 1024 | 83 | ||
BC012187.1 | - | 8 - 9 | 1162 | 4634631 | 83 | |
CR456448.1 | - | 8 - 9 | 918 | 83 | ||
KU178842.1 | - | 8 - 9 | 1011 | 83 | ||
KU178843.1 | - | 8 - 9 | 885 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |