Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015190.4 | NP_056005.1 | 1 - 2 | 322 | 107 | |
GenBank mRNA | AF327347.1 | - | 1 - 2 | 229 | 107 | |
AK074420.1 | - | 1 - 2 | 258 | 107 | ||
AK094162.1 | - | 1 - 2 | 1853 | 107 | ||
BC136507.1 | - | 1 - 2 | 249 | 9020496 | 107 | |
BX646217.1 | - | 1 - 2 | 322 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |