Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256508.1 | NP_001243437.1 | 5 - 6 | 508 | 86 | |
XM_006718541.3 | XP_006718604.1 | 5 - 6 | 792 | 86 | ||
XM_006718542.3 | XP_006718605.1 | 5 - 6 | 775 | 86 | ||
XM_006718543.2 | XP_006718606.1 | 5 - 6 | 509 | 86 | ||
XM_006718544.3 | XP_006718607.1 | 5 - 6 | 703 | 86 | ||
XM_011545001.2 | XP_011543303.1 | 4 - 5 | 730 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |