Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021334.hg.1 |
ENST00000315286.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021334.hg.1 |
ENST00000457422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021334.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021334.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021334.hg.1 |
TC17001584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000588687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000588687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010841.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000315286.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000457422.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000588210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
ENST00000592333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
NM_001099225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
NM_144609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186806.hg.1 |
TC1700010841.hg.1 |
Clariom D TC |