Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000587478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000587478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000587478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006751.hg.1 |
ENST00000445254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006751.hg.1 |
ENST00000445254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006826.hg.1 |
ENST00000480072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000587478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000587478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000587478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007930.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006826.hg.1 |
ENST00000480072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000253788.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000586277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000588830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000589037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000589913.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000590864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
ENST00000593262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
NM_000988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157945.hg.1 |
TC1700007930.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006751.hg.1 |
ENST00000445254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006751.hg.1 |
ENST00000445254.1 |
Ensembl |