Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SNORD94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_004378.1 | 1 | 72 | 76 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pentatricopeptide repeat domain 3 | PTCD3 | MRP-S39 | Sense | Chr. 2 - 86106182 - 86142157 on NCBI Build GRCh38 | |
Downstream | inner membrane mitochondrial protein | IMMT |
HMP, MINOS2, Mic60, P87, P87/89, P89, PIG4, PIG52 |
7.9kb | Anti-sense | Chr. 2 - 86143932 - 86195770 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4295 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4296 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415996 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333269 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415939 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415929 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333296 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415942 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415991 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333310 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415977 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415986 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415950 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333286 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333284 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415930 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415932 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415951 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415972 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415981 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415999 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415999 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415972 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333284 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415981 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333296 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415929 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415950 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415986 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415996 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415930 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333310 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415977 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415951 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415932 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333269 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415939 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415991 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333286 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21415942 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01333252 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333271 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333249 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333278 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333243 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333293 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333246 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333241 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333264 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333307 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333266 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333314 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333283 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333305 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333253 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333309 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333290 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333255 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333301 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333247 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333297 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333240 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333269 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333312 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333273 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333288 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333280 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333276 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333257 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333285 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01333262 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15264067 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_004378_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043277 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8043276 | - | - |
Affymetrix Human Genome U219 Array | 11757173_x_at | - | - |
Affymetrix Human Genome U219 Array | 11757173_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0200008283.hg.1 | ENST00000386037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008283.hg.1 | NR_004378.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02008124.hg.1 | TC02000521.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02008124.hg.1 | ENST00000386037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02008124.hg.1 | NR_004378.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008283.hg.1 | ENST00000386037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008283.hg.1 | NR_004378.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157556.hg.1 | ENST00000386037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157556.hg.1 | NR_004378.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157556.hg.1 | ENST00000386037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157556.hg.1 | NR_004378.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157556.hg.1 | TC0200008283.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3245274 | NR_004378.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3245274 | NR_004378.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245274 | NR_004378.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245274 | NR_004378.1 | human_transcriptome |