Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY927437.1 | - | LIT3470 AY927437 |
7 - 8 | 624 | 123 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ring finger protein 215 | RNF215 | -35.0kb | Anti-sense | Chr. 22 - 30378812 - 30387313 on NCBI Build GRCh38 | |
Downstream | mitochondrial fission process 1 | MTFP1 |
HSPC242, MTP18 |
686.0 | Sense | Chr. 22 - 30425623 - 30429054 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |