Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, H/ACA box 70 |
SNORA70 |
DXS648E, RNU70, U70 |
-49.0 | Anti-sense | Chr. X - 154400281 - 154400415 on NCBI Build GRCh38 |
Downstream | tafazzin | TAZ |
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1 |
8.0kb | Anti-sense | Chr. X - 154411406 - 154421726 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |