Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 245 | TMEM245 |
C9orf5, CG-2, CG2 |
Sense | Chr. 9 - 109015135 - 109119963 on NCBI Build GRCh38 | |
Downstream | microRNA 32 | MIR32 |
MIRN32, hsa-mir-32, miR-32, miRNA32 |
1.5kb | Sense | Chr. 9 - 109046229 - 109046298 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |