Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
ENST00000489671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
ENST00000499676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
ENST00000602556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_006286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
ENST00000489671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
ENST00000499676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
ENST00000602556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_001178142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
NM_006286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194458.hg.1 |
TC0300012654.hg.1 |
Clariom D TC |