Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7927366 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927365 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927366 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933328 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928904 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928906 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933424 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927364 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928905 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933329 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928905 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933330 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933426 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933424 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928904 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927365 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933425 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933425 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933330 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927364 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927364 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933424 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928905 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933329 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933426 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933426 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928906 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927365 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933329 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933328 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933330 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933425 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7927366 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928904 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7928906 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7933328 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7927363 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7927363 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7928890 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7933327 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7927363 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7933327 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7933327 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7933423 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7933423 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7933423 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7928890 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7928890 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015474.hg.1 |
ENST00000340853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015474.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015474.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015474.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015474.hg.1 |
TC10001239.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000608562.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010507.hg.1 |
ENST00000608562.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012552.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012554.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012554.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012554.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012554.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012554.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012554.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187157.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187157.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187157.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187157.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187157.hg.1 |
TC1000012554.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185586.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185586.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185586.hg.1 |
TC1000012453.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185825.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185825.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185825.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185825.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185825.hg.1 |
TC1000012472.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185587.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185587.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185587.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185587.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185587.hg.1 |
TC1000012453.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185593.hg.1 |
TC1000012453.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185594.hg.1 |
TC1000012453.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187134.hg.1 |
TC1000012552.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000411791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000426412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000608562.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010507.hg.1 |
ENST00000608562.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185592.hg.1 |
TC1000012453.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
ENST00000343959.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012472.hg.1 |
NM_001146157.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
ENST00000452267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
ENST00000481177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012552.hg.1 |
NM_001137549.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012554.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012554.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012554.hg.1 |
ENST00000617224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012554.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012554.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012554.hg.1 |
NM_001137548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000623997.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
ENST00000624613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012453.hg.1 |
NR_104039.1 |
Refseq |
Human Genome U95 Set |
57452_at |
- |
- |
Human Genome U95 Set |
57452_PM_at |
- |
- |
Human Genome U95 Set |
57452_at |
- |
- |
Human Genome U95 Set |
57452_PM_at |
- |
- |
Human Genome U95 Set |
57452_at |
- |
- |
Human Genome U95 Set |
57452_PM_at |
- |
- |