Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000551.3 | NP_000542.1 | 1 | 56 | 78 | |
NM_198156.2 | NP_937799.1 | 1 | 56 | 78 | ||
GenBank mRNA | L15409.1 | - | 1 | 56 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |