Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244962.1 | NP_001231891.1 | 1 - 2 | 282 | 73 | |
XM_017014591.1 | XP_016870080.1 | 1 - 2 | 294 | 73 | ||
GenBank mRNA | BC023560.1 | - | 1 - 2 | 282 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |